Please login to your online account to display your discounted pricing

2-Amino-6-purinethiol, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  104480010

 View more versions of this product

Catalog No. AC104480010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Color Yellow to green
Formula Weight 167.19
Physical Form Powder
Quantity 1g
Beilstein 26, IV, 3926
Merck Index 15, 9490
CAS Max % 100.0
CAS Min % 94.0
Chemical Name or Material 2-Amino-6-purinethiol
Assay Percent Range 95%
Health Hazard 1 Danger
Packaging Glass bottle
Synonym 6-Thioguanine
Infrared Spectrum Authentic
MDL Number MFCD00127785
Molecular Formula C5H5N5S
CAS 154-42-7
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Amino-6-purinethiol
CAS 154-42-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H5N5S
Formula Weight 167.19
MDL Number MFCD00127785
Synonym 6-Thioguanine
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.

SDS & Safety Documents