Please login to your online account to display your discounted pricing

2-Amino-6-picoline 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  104405000

 View more versions of this product

Catalog No. AC104405000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Low Melting Solid
Formula Weight 108.14
Quantity 500g
Color Yellow
EINECS Number 217-360-1
TSCA TSCA
RTECS Number US1885000
Melting Point 40° to 45°C
CAS Max % 100.0
Beilstein 22, V,9, 210
Synonym 2-Amino-6-methylpyridine
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00006331
Chemical Name or Material 2-Amino-6-picoline
Name Note 98%
CAS 1824-81-3
CAS Min % 97.5
Assay Percent Range 98%
Molecular Formula C6H8N2
Packaging Glass bottle
Water 0.5% max.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2-Amino-6-picoline
Name Note 98%
CAS 1824-81-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H8N2
Formula Weight 108.14
MDL Number MFCD00006331
Synonym 2-Amino-6-methylpyridine
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.

SDS & Safety Documents