missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-picoline 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 104401000

 View more versions of this product

Catalog No. AC104401000


 

Specifications Description & Specifications

Specifications

≥97.5%
Crystalline Low Melting Solid
Yellow
100.0
98%
97.5
1824-81-3
98%
Glass bottle
Authentic
108.14
100g
40° to 45°C
22, V,9, 210
MFCD00006331
2-Amino-6-picoline
C6H8N2
0.5% max.
2-Amino-6-methylpyridine

Chemical Identifiers

1824-81-3
100.0
C6H8N2
MFCD00006331
2-Amino-6-picoline, 98%
97.5
98%
108.14
2-Amino-6-methylpyridine
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 217-360-1

RTECSNumber : US1885000

TSCA : TSCA