missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-nitrobenzoic acid, 97%

Manufacturer:  Alfa Aesar™ H66090MD

 View more versions of this product

Catalog No. AAH66090MD



Specifications

Specifications

250mg
C7H6N2O4
2807130
GGKYLHNARFFORH-UHFFFAOYSA-N
182.135
182.13
97%
50573-74-5
MFCD00034785
2-amino-6-nitro-benzoic acid, 2-amino-6-nitrobenzoic acid, 2-amino-6-nitrobenzoicacid, 6-nitro-2-aminobenzoesaure, 6-nitro-anthranilic acid, 6-nitroanthanilic acid, 6-nitroanthranilic acid, acmc-209knw, benzoic acid, 2-amino-6-nitro, ksc911o8d
C1=CC(=C(C(=C1)[N+](=O)[O-])C(=O)O)N
95795
Solid

Chemical Identifiers

50573-74-5
182.135
GGKYLHNARFFORH-UHFFFAOYSA-N
95795
C7H6N2O4
MFCD00034785
2-amino-6-nitro-benzoic acid, 2-amino-6-nitrobenzoic acid, 2-amino-6-nitrobenzoicacid, 6-nitro-2-aminobenzoesaure, 6-nitro-anthranilic acid, 6-nitroanthanilic acid, 6-nitroanthranilic acid, acmc-209knw, benzoic acid, 2-amino-6-nitro, ksc911o8d
C1=CC(=C(C(=C1)[N+](=O)[O-])C(=O)O)N
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 610-543-0

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents