missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-nitrobenzoic acid, 97%

Manufacturer:  Alfa Aesar™ H6609006

 View more versions of this product

Catalog No. AAH6609006



Specifications

Specifications

2-Amino-6-nitrobenzoic acid
50573-74-5
MFCD00034785
2-amino-6-nitrobenzoic acid, benzoic acid, 2-amino-6-nitro, 6-nitroanthranilic acid, 2-amino-6-nitro-benzoic acid, 2-amino-6-nitrobenzoicacid, 6-nitroanthanilic acid, acmc-209knw, 6-nitro-anthranilic acid, 6-nitro-2-aminobenzoesaure, ksc911o8d
C1=CC(=C(C(=C1)[N+](=O)[O-])C(=O)O)N
95795
Solid
5g
C7H6N2O4
2807130
GGKYLHNARFFORH-UHFFFAOYSA-N
182.135
182.13
97%

Chemical Identifiers

50573-74-5
182.135
GGKYLHNARFFORH-UHFFFAOYSA-N
95795
C7H6N2O4
MFCD00034785
2-amino-6-nitrobenzoic acid, benzoic acid, 2-amino-6-nitro, 6-nitroanthranilic acid, 2-amino-6-nitro-benzoic acid, 2-amino-6-nitrobenzoicacid, 6-nitroanthanilic acid, acmc-209knw, 6-nitro-anthranilic acid, 6-nitro-2-aminobenzoesaure, ksc911o8d
C1=CC(=C(C(=C1)[N+](=O)[O-])C(=O)O)N
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

Documents