missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-methylpyrazine, 97%

$101.32 - $303.80

Chemical Identifiers

CAS 5521-56-2
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD08235193
InChI Key UAOOJJPSCLNTOP-UHFFFAOYSA-N
Synonym 2-amino-6-methylpyrazine, pyrazinamine, 6-methyl, 6-methylpyrazine-2-ylamine, 6-methyl-2-pyrazinamine, 2-pyrazinamine,6-methyl, 2-amino-6-methyl pyrazine, 6-methyl-pyrazin-2-ylamine, acmc-1am75, 6-methyl-pyrazinamine, 6-methylpyrazin-2-ylamine
PubChem CID 12236939
IUPAC Name 6-methylpyrazin-2-amine
SMILES CC1=CN=CC(=N1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63436MD View Documents Alfa Aesar™
H63436MD
250mg Each for $101.32
 
AAH6343603 View Documents Alfa Aesar™
H6343603
1g Each for $303.80
 
Specifications

Chemical Identifiers

5521-56-2
109.132
UAOOJJPSCLNTOP-UHFFFAOYSA-N
12236939
CC1=CN=CC(=N1)N
C5H7N3
MFCD08235193
2-amino-6-methylpyrazine, pyrazinamine, 6-methyl, 6-methylpyrazine-2-ylamine, 6-methyl-2-pyrazinamine, 2-pyrazinamine,6-methyl, 2-amino-6-methyl pyrazine, 6-methyl-pyrazin-2-ylamine, acmc-1am75, 6-methyl-pyrazinamine, 6-methylpyrazin-2-ylamine
6-methylpyrazin-2-amine

Specifications

250mg
C5H7N3
2-amino-6-methylpyrazine, pyrazinamine, 6-methyl, 6-methylpyrazine-2-ylamine, 6-methyl-2-pyrazinamine, 2-pyrazinamine,6-methyl, 2-amino-6-methyl pyrazine, 6-methyl-pyrazin-2-ylamine, acmc-1am75, 6-methyl-pyrazinamine, 6-methylpyrazin-2-ylamine
CC1=CN=CC(=N1)N
109.132
109.13
97%
5521-56-2
MFCD08235193
UAOOJJPSCLNTOP-UHFFFAOYSA-N
6-methylpyrazin-2-amine
12236939
Solid
2-Amino-6-methylpyrazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning