Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-methoxypyrazine, 95%

Chemical Identifiers

Chemical Name or Material 2-Amino-6-methoxypyrazine
CAS 6905-47-1
Formula Weight 125.13
MDL Number MFCD09909689
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6199003

  SDS
alfa aesar™
H6199003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61990MD

  SDS
alfa aesar™
H61990MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 125.13
CAS 6905-47-1
Assay 95%
Chemical Name or Material 2-Amino-6-methoxypyrazine
Quantity 1g
Physical Form Solid
MDL Number MFCD09909689