missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-6-methoxybenzothiazole, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 172020500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Amino-6-methoxybenzothiazole | |
1747-60-0 | |
100.0 | |
98% | |
C8H8N2OS | |
50g | |
2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393 | |
COC1=CC2=C(C=C1)N=C(S2)N | |
180.23 | |
180.23 |
98% | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00005787 | |
27, II, 334 | |
KZHGPDSVHSDCMX-UHFFFAOYSA-N | |
6-methoxy-1,3-benzothiazol-2-amine | |
15630 | |
98% |
Chemical Identifiers
1747-60-0 | |
180.23 | |
KZHGPDSVHSDCMX-UHFFFAOYSA-N | |
15630 | |
COC1=CC2=C(C=C1)N=C(S2)N |
C8H8N2OS | |
MFCD00005787 | |
2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393 | |
6-methoxy-1,3-benzothiazol-2-amine |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Danger
EINECSNumber : 217-130-0
RTECSNumber : DL2100000
TSCA : TSCA
RUO â Research Use Only