Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-methoxy-3-nitropyridine, 97%

$83.49 - $629.82

Chemical Identifiers

Chemical Name or Material 2-Amino-6-methoxy-3-nitropyridine
CAS 73896-36-3
Molecular Formula C6H7N3O3
Formula Weight 169.14
MDL Number MFCD03206481
Synonym 6-Methoxy-3-nitro-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2666803

  SDS
alfa aesar™
H2666803
1g Each for $207.93

AAH2666806

  SDS
alfa aesar™
H2666806
5g Each for $629.82

AAH26668MD

  SDS
alfa aesar™
H26668MD
250mg Each for $83.49
Description & Specifications

Specifications

Formula Weight 169.14
Chemical Name or Material 2-Amino-6-methoxy-3-nitropyridine
Assay 97%
Molecular Formula C6H7N3O3
Melting Point 170° to 172°C
MDL Number MFCD03206481
CAS 73896-36-3
Quantity 1g
Synonym 6-Methoxy-3-nitro-2-pyridinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.