Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-iodopurine, 97%

$35.41 - $132.07

Chemical Identifiers

Chemical Name or Material 2-Amino-6-iodopurine
CAS 19690-23-4
Molecular Formula C5H4IN5
Formula Weight 261.02
MDL Number MFCD08706375
Synonym 6-Iodo-9H-purin-2-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5167203

 
alfa aesar™
H5167203
1g Each for $35.41

AAH5167206

 
alfa aesar™
H5167206
5g Each for $132.07
Description & Specifications

Specifications

Formula Weight 261.02
Chemical Name or Material 2-Amino-6-iodopurine
Assay 97%
Molecular Formula C5H4IN5
MDL Number MFCD08706375
CAS 19690-23-4
Quantity 1g
Synonym 6-Iodo-9H-purin-2-amine