missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-hydroxy-8-mercaptopurine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 295230050

 View more versions of this product

Catalog No. AC295230050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-6-hydroxy-8-mercaptopurine
Glass bottle
>300.0°C
97%
96.0
97%
MFCD00075623
JHEKNTQSGTVPAO-UHFFFAOYSA-N
2-amino-8-sulfanylidene-7,9-dihydro-3H-purin-6-one
2725005
Fine Crystalline Powder
Authentic
Yellow
5g
28128-40-7
100.0
C5H5N5OS
2-amino-1,7,8,9-tetrahydro-2-thioxo-6h-purin-6-one, 2-amino-6-hydroxy-8-mercaptopurine, 2-amino-8-mercapto-1,9-dihydro-purin-6-one, 2-amino-8-mercapto-7h-purin-6-ol, 2-amino-8-sulfanyl-1,9-dihydro-6h-purin-6-one, 2-amino-8-sulfanyl-7h-purin-6-ol, 2-amino-8-sulfanyl-9h-purin-6-ol, 2-amino-8-thioxo-1,7,8,9-tetrahydro-purin-6-one, 2-amino-8-thioxo-3,7-dihydropurin-6-one, 8-mercaptoguanine
C12=C(NC(=NC1=O)N)NC(=S)N2
183.19
183.19
97%

Chemical Identifiers

28128-40-7
183.19
JHEKNTQSGTVPAO-UHFFFAOYSA-N
2725005
C12=C(NC(=NC1=O)N)NC(=S)N2
C5H5N5OS
MFCD00075623
2-amino-1,7,8,9-tetrahydro-2-thioxo-6h-purin-6-one, 2-amino-6-hydroxy-8-mercaptopurine, 2-amino-8-mercapto-1,9-dihydro-purin-6-one, 2-amino-8-mercapto-7h-purin-6-ol, 2-amino-8-sulfanyl-1,9-dihydro-6h-purin-6-one, 2-amino-8-sulfanyl-7h-purin-6-ol, 2-amino-8-sulfanyl-9h-purin-6-ol, 2-amino-8-thioxo-1,7,8,9-tetrahydro-purin-6-one, 2-amino-8-thioxo-3,7-dihydropurin-6-one, 8-mercaptoguanine
2-amino-8-sulfanylidene-7,9-dihydro-3H-purin-6-one
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents