Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-hydroxy-8-mercaptopurine, 97%

$60.66 - $192.41

Chemical Identifiers

Chemical Name or Material 2-Amino-6-hydroxy-8-mercaptopurine
CAS 28128-40-7
Molecular Formula C5H5N5OS
Formula Weight 183.19
MDL Number MFCD00075623
Beilstein 982327
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0777703

 
alfa aesar™
L0777703
1g Each for $60.66

AAL0777706

 
alfa aesar™
L0777706
5g Each for $192.41
Description & Specifications

Specifications

Formula Weight 183.19
Chemical Name or Material 2-Amino-6-hydroxy-8-mercaptopurine
Assay 97%
Molecular Formula C5H5N5OS
MDL Number MFCD00075623
Quantity 1g
Melting Point >300°C
CAS 28128-40-7
Beilstein 982327