Please login to your online account to display your discounted pricing

2-Amino-6-fluorobenzothiazole, 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  263730100

 View more versions of this product

Catalog No. AC263730100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Additional Information product discolors on exposure to light
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 181° to 185°C
Color Off-white to tan
Quantity 10g
Chemical Name or Material 2-Amino-6-fluorobenzothiazole
Name Note 99%
CAS 348-40-3
CAS Min % 98.5
CAS Max % 100.0
Physical Form Off-white to tan crystalline powder
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C7H5FN2S
Formula Weight 168.20g/mol
MDL Number MFCD00013336
Synonym 6-Fluoro-2-benzothiazolamine
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

Specifications

Chemical Name or Material 2-Amino-6-fluorobenzothiazole
Name Note 99%
CAS 348-40-3
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C7H5FN2S
Formula Weight 168.20g/mol
MDL Number MFCD00013336
Synonym 6-Fluoro-2-benzothiazolamine
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents