missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-fluorobenzothiazole, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros AC263730010

 View more versions of this product

Catalog No. AC263730010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Amino-6-fluorobenzothiazole
Glass bottle
181.0°C to 185.0°C
99%
98.5
98.5% min. (GC)
MFCD00013336
CJLUXPZQUXVJNF-UHFFFAOYSA-N
6-fluoro-1,3-benzothiazol-2-amine
319954
Crystalline Powder
Authentic
Tan to White
1g
348-40-3
100.0
C7H5FN2S
2-amino-6-fluoro benzothiazole, 2-amino-6-fluoro-1,3-benzothiazole, 2-amino-6-fluorobenzothiazole, 2-benzothiazolamine, 6-fluoro, 6-fluoro-2-benzothiazolamine, 6-fluoro-benzothiazol-2-ylamine, 6-fluorobenzo d thiazol-2-amine, 6-fluorobenzothiazole-2-ylamine, maybridge1_006546, pubchem14467
C1=CC2=C(C=C1F)SC(=N2)N
168.2
168.2
99%

Chemical Identifiers

348-40-3
168.2
CJLUXPZQUXVJNF-UHFFFAOYSA-N
319954
C1=CC2=C(C=C1F)SC(=N2)N
C7H5FN2S
MFCD00013336
2-amino-6-fluoro benzothiazole, 2-amino-6-fluoro-1,3-benzothiazole, 2-amino-6-fluorobenzothiazole, 2-benzothiazolamine, 6-fluoro, 6-fluoro-2-benzothiazolamine, 6-fluoro-benzothiazol-2-ylamine, 6-fluorobenzo d thiazol-2-amine, 6-fluorobenzothiazole-2-ylamine, maybridge1_006546, pubchem14467
6-fluoro-1,3-benzothiazol-2-amine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents