Learn More
2-Amino-6-fluorobenzothiazole, 99%, ACROS Organics™
Chemical Identifiers
CAS | 348-40-3 |
---|---|
Molecular Formula | C7H5FN2S |
Molecular Weight (g/mol) | 168.2 |
MDL Number | MFCD00013336 |
InChI Key | CJLUXPZQUXVJNF-UHFFFAOYSA-N |
Synonym | 2-amino-6-fluoro benzothiazole, 2-amino-6-fluoro-1,3-benzothiazole, 2-amino-6-fluorobenzothiazole, 2-benzothiazolamine, 6-fluoro, 6-fluoro-2-benzothiazolamine, 6-fluoro-benzothiazol-2-ylamine, 6-fluorobenzo d thiazol-2-amine, 6-fluorobenzothiazole-2-ylamine, maybridge1_006546, pubchem14467 |
PubChem CID | 319954 |
IUPAC Name | 6-fluoro-1,3-benzothiazol-2-amine |
SMILES | C1=CC2=C(C=C1F)SC(=N2)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC263730010
|
Fisher Scientific Acros
AC263730010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
348-40-3 | |
168.2 | |
CJLUXPZQUXVJNF-UHFFFAOYSA-N | |
319954 | |
C1=CC2=C(C=C1F)SC(=N2)N |
C7H5FN2S | |
MFCD00013336 | |
2-amino-6-fluoro benzothiazole, 2-amino-6-fluoro-1,3-benzothiazole, 2-amino-6-fluorobenzothiazole, 2-benzothiazolamine, 6-fluoro, 6-fluoro-2-benzothiazolamine, 6-fluoro-benzothiazol-2-ylamine, 6-fluorobenzo d thiazol-2-amine, 6-fluorobenzothiazole-2-ylamine, maybridge1_006546, pubchem14467 | |
6-fluoro-1,3-benzothiazol-2-amine |
Specifications
Authentic | |
Tan to White | |
1g | |
98.5 | |
98.5% min. (GC) | |
MFCD00013336 | |
CJLUXPZQUXVJNF-UHFFFAOYSA-N | |
6-fluoro-1,3-benzothiazol-2-amine | |
319954 | |
Crystalline Powder | |
2-Amino-6-fluorobenzothiazole, 99% |
Glass bottle | |
181.0°C to 185.0°C | |
348-40-3 | |
100.0 | |
C7H5FN2S | |
2-amino-6-fluoro benzothiazole, 2-amino-6-fluoro-1,3-benzothiazole, 2-amino-6-fluorobenzothiazole, 2-benzothiazolamine, 6-fluoro, 6-fluoro-2-benzothiazolamine, 6-fluoro-benzothiazol-2-ylamine, 6-fluorobenzo d thiazol-2-amine, 6-fluorobenzothiazole-2-ylamine, maybridge1_006546, pubchem14467 | |
C1=CC2=C(C=C1F)SC(=N2)N | |
168.2 | |
168.2 | |
99% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA