Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-fluorobenzonitrile, 99%

Chemical Identifiers

Chemical Name or Material 2-Amino-6-fluorobenzonitrile
CAS 77326-36-4
Molecular Formula C7H5FN2
Formula Weight 136.13
MDL Number MFCD00015477
UN Number UN3439
Beilstein 3588945
Synonym 6-Fluoroanthranilonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1245603

  SDS
alfa aesar™
A1245603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1245606

  SDS
alfa aesar™
A1245606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 136.13
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 6-Fluoroanthranilonitrile
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD00015477
Health Hazard 1 Danger
Melting Point 125° to 129°C
Quantity 1g
Assay 99%
Molecular Formula C7H5FN2
CAS 77326-36-4
Chemical Name or Material 2-Amino-6-fluorobenzonitrile
UN Number UN3439
Beilstein 3588945
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.