Please login to your online account to display your discounted pricing

2-Amino-6-fluorobenzoic acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  306450010

 View more versions of this product

Catalog No. AC306450010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 155.13
Quantity 1g
Color Yellow to brown
Melting Point 165° to 169°C
CAS Max % 100.0
Linear Formula HNC6H3(F)COOH
Synonym 6-Fluoroanthranilic acid
Molecular Formula C7H6FNO2
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00067781
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 434-76-4
Chemical Name or Material 2-Amino-6-fluorobenzoic acid
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material 2-Amino-6-fluorobenzoic acid
CAS 434-76-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H6FNO2
Linear Formula HNC6H3(F)COOH
Formula Weight 155.13
MDL Number MFCD00067781
Synonym 6-Fluoroanthranilic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents