Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-ethylpyridine, 97%

$32.22 - $32.22

Chemical Identifiers

Chemical Name or Material 2-Amino-6-ethylpyridine
CAS 21717-29-3
Molecular Formula C7H10N2
Formula Weight 122.17
MDL Number MFCD00218265
UN Number UN2810
Synonym 6-Ethyl-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1914618

  SDS
alfa aesar™
A1914618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1914609

  SDS
alfa aesar™
A1914609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1914688

  SDS
alfa aesar™
A1914688
2.5g Each for $32.22
Description & Specifications

Specifications

Formula Weight 122.17
Chemical Name or Material 2-Amino-6-ethylpyridine
Assay 97%
Quantity 50g
Odor Unpleasant
Molecular Formula C7H10N2
MDL Number MFCD00218265
CAS 21717-29-3
UN Number UN2810
Synonym 6-Ethyl-2-pyridinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Applications
The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.