missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-ethoxybenzothiazole 97%, ACROS Organics™

Manufacturer:  Acros Organics 103621000

 View more versions of this product

Catalog No. AC103621000



Specifications

Specifications

2-Amino-6-ethoxybenzothiazole
Glass bottle
Colorless to Yellow
100g
94-45-1
100.0
C9H10N2OS
2-amino-6-ethoxybenzothiazole
CCOC1=CC2=C(C=C1)N=C(S2)N
194.252
194.25
≥96.0% (total base)
Authentic
1% max.
160°C to 166°C
97%
96.0
97%
MFCD00005788
KOYJWFGMEBETBU-UHFFFAOYSA-N
6-ethoxy-1,3-benzothiazol-2-amine
7192
Liquid

Chemical Identifiers

94-45-1
194.252
KOYJWFGMEBETBU-UHFFFAOYSA-N
7192
CCOC1=CC2=C(C=C1)N=C(S2)N
C9H10N2OS
MFCD00005788
2-amino-6-ethoxybenzothiazole
6-ethoxy-1,3-benzothiazol-2-amine
Safety and Handling

Safety and Handling

EINECSNumber : 202-333-9

SDS Documents