Please login to your online account to display your discounted pricing

2-Amino-6-ethoxybenzothiazole 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  103620250

 View more versions of this product

Catalog No. AC103620250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (total base)
Physical Form Liquid
Formula Weight 194.25
Quantity 25g
Color Clear colorless to yellow
EINECS Number 202-333-9
Melting Point 160° to 166°C
CAS Max % 100.0
CAS Min % 96.0
Chemical Name or Material 2-Amino-6-ethoxybenzothiazole
Name Note 97%
Molecular Formula C9H10N2OS
MDL Number MFCD00005788
CAS 94-45-1
Infrared Spectrum Authentic
Water 1% max.
Assay Percent Range 97%
Packaging Glass bottle

Specifications

Chemical Name or Material 2-Amino-6-ethoxybenzothiazole
Name Note 97%
CAS 94-45-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (total base)
Assay Percent Range 97%
Molecular Formula C9H10N2OS
Formula Weight 194.25
MDL Number MFCD00005788