missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-6-ethoxybenzothiazole 97%, ACROS Organics™
$183.74 - $183.74
Chemical Identifiers
CAS | 94-45-1 |
---|---|
Molecular Formula | C9H10N2OS |
Molecular Weight (g/mol) | 194.252 |
MDL Number | MFCD00005788 |
InChI Key | KOYJWFGMEBETBU-UHFFFAOYSA-N |
Synonym | 2-amino-6-ethoxybenzothiazole, 2-benzothiazolamine, 6-ethoxy, 2-benzothiazolamine, 6-ethoxy-9ci, 6-ethoxy-2-aminobenzothiazole, 6-ethoxy-2-benzothiazolamine, 6-ethoxy-benzothiazol-2-ylamine, 6-ethoxybenzothiazol-2-ylamine, amino-6-ethoxybenzothiazole, 2, benzothiazole, 2-amino-6-ethoxy, unii-d621hz191d |
PubChem CID | 7192 |
IUPAC Name | 6-ethoxy-1,3-benzothiazol-2-amine |
SMILES | CCOC1=CC2=C(C=C1)N=C(S2)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC103621000
|
Thermo Fisher Scientific
103621000 |
100g | Glass bottle |
Each for $183.74
|
|
Chemical Identifiers
94-45-1 | |
194.252 | |
KOYJWFGMEBETBU-UHFFFAOYSA-N | |
7192 | |
CCOC1=CC2=C(C=C1)N=C(S2)N |
C9H10N2OS | |
MFCD00005788 | |
2-amino-6-ethoxybenzothiazole, 2-benzothiazolamine, 6-ethoxy, 2-benzothiazolamine, 6-ethoxy-9ci, 6-ethoxy-2-aminobenzothiazole, 6-ethoxy-2-benzothiazolamine, 6-ethoxy-benzothiazol-2-ylamine, 6-ethoxybenzothiazol-2-ylamine, amino-6-ethoxybenzothiazole, 2, benzothiazole, 2-amino-6-ethoxy, unii-d621hz191d | |
6-ethoxy-1,3-benzothiazol-2-amine |
Specifications
Authentic | |
1% max. | |
160°C to 166°C | |
94-45-1 | |
100.0 | |
C9H10N2OS | |
2-amino-6-ethoxybenzothiazole, 2-benzothiazolamine, 6-ethoxy, 2-benzothiazolamine, 6-ethoxy-9ci, 6-ethoxy-2-aminobenzothiazole, 6-ethoxy-2-benzothiazolamine, 6-ethoxy-benzothiazol-2-ylamine, 6-ethoxybenzothiazol-2-ylamine, amino-6-ethoxybenzothiazole, 2, benzothiazole, 2-amino-6-ethoxy, unii-d621hz191d | |
CCOC1=CC2=C(C=C1)N=C(S2)N | |
194.252 | |
194.25 | |
≥96.0% (total base) |
Glass bottle | |
Colorless to Yellow | |
100g | |
96.0 | |
97% | |
MFCD00005788 | |
KOYJWFGMEBETBU-UHFFFAOYSA-N | |
6-ethoxy-1,3-benzothiazol-2-amine | |
7192 | |
Liquid | |
2-Amino-6-ethoxybenzothiazole, 97% |
Safety and Handling
EINECSNumber : 202-333-9