Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-ethoxybenzothiazole, 97%

$48.80 - $128.53

Chemical Identifiers

Chemical Name or Material 2-Amino-6-ethoxybenzothiazole
CAS 94-45-1
Molecular Formula C9H10N2OS
Formula Weight 194.25
MDL Number MFCD00005788
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2376114

  SDS
alfa aesar™
B2376114
25g Each for $48.80

AAB2376122

  SDS
alfa aesar™
B2376122
100g Each for $128.53
Description & Specifications

Specifications

Formula Weight 194.25
Chemical Name or Material 2-Amino-6-ethoxybenzothiazole
Melting Point 161° to 163°C
Assay 97%
Quantity 25g
Molecular Formula C9H10N2OS
MDL Number MFCD00005788
CAS 94-45-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.