missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-chloropurine riboside, 95%

$147.73 - $294.54

Chemical Identifiers

CAS 2004-07-1
Molecular Formula C10H12ClN5O4
Molecular Weight (g/mol) 301.69
MDL Number MFCD00005735
InChI Key TXWHPSZYRUHEGT-UHFFFAOYNA-N
Synonym 2-amino-6-chloropurine riboside, 2-amino-6-chloropurine-9-riboside
PubChem CID 46874491
IUPAC Name (3R,4S,5R)-2-(2-amino-6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2Cl)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6446803 View Documents Alfa Aesar™
J6446803
1g Each for $147.73
 
AAJ6446806 View Documents Alfa Aesar™
J6446806
5g Each for $294.54
 
Specifications

Chemical Identifiers

2004-07-1
301.69
TXWHPSZYRUHEGT-UHFFFAOYNA-N
46874491
C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2Cl)N
C10H12ClN5O4
MFCD00005735
2-amino-6-chloropurine riboside, 2-amino-6-chloropurine-9-riboside
(3R,4S,5R)-2-(2-amino-6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol

Specifications

White
1g
C10H12ClN5O4
2-amino-6-chloropurine riboside, 2-amino-6-chloropurine-9-riboside
C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2Cl)N
301.69
301.69
95%
165°C to 167°C
2004-07-1
MFCD00005735
TXWHPSZYRUHEGT-UHFFFAOYNA-N
(3R,4S,5R)-2-(2-amino-6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
46874491
Powder
2-Amino-6-chloropurine riboside
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C