Learn More
2-Amino-6-chloropurine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1819514
Description
2-Amino-6-chloropurine finds its uses in the enzymatic synthesis of 2?-deoxyguanosine, 9-alkyl purines and (R)- and (S)-N-(2-phosphonomethoxypropyl) derivatives of purine and pyrimidine bases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-6-chloropurine finds its uses in the enzymatic synthesis of 2′-deoxyguanosine, 9-alkyl purines and (R)- and (S)-N-(2-phosphonomethoxypropyl) derivatives of purine and pyrimidine bases.
Solubility
Solubility in 1mol/L NaOH; very faint turbidity.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
2-Amino-6-chloropurine | |
>300°C | |
MFCD00075252 | |
9626 | |
RYYIULNRIVUMTQ-UHFFFAOYSA-N | |
6-chloro-7H-purin-2-amine | |
5360349 | |
169.57 |
10310-21-1 | |
C5H4ClN5 | |
25 g | |
2-amino-6-chloropurine, 6-chloroguanine, 6-chloro-9h-purin-2-amine, 6-chloro-2-aminopurine, 2-amino-6-chlorpurine, 1h-purin-2-amine, 6-chloro, purine, 2-amino-6-chloro, 6-chloro-7h-purin-2-ylamine, 2-amino-6-chloro-purin, 2-amino-6-chloro-purine | |
NC1=NC(Cl)=C2NC=NC2=N1 | |
169.57 | |
CHEBI:72345 | |
99% |
Chemical Identifiers
10310-21-1 | |
169.57 | |
RYYIULNRIVUMTQ-UHFFFAOYSA-N | |
5360349 | |
6-chloro-7H-purin-2-amine |
C5H4ClN5 | |
MFCD00075252 | |
2-amino-6-chloropurine, 6-chloroguanine, 6-chloro-9h-purin-2-amine, 6-chloro-2-aminopurine, 2-amino-6-chlorpurine, 1h-purin-2-amine, 6-chloro, purine, 2-amino-6-chloro, 6-chloro-7h-purin-2-ylamine, 2-amino-6-chloro-purin, 2-amino-6-chloro-purine | |
CHEBI:72345 | |
NC1=NC(Cl)=C2NC=NC2=N1 |
Safety and Handling
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H360
EINECSNumber : 233-686-7
RTECSNumber : UO7502000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only