missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-chlorobenzothiazole, 99%, ACROS Organics™

Chemical Identifiers

CAS 95-24-9
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C7H5ClN2S
Formula Weight 184.65
MDL Number MFCD00053557
Chemical Name or Material 2-Amino-6-chlorobenzothiazole
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC136400500 SDS Thermo Fisher Scientific
AC136400500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

95-24-9
100.0
C7H5ClN2S
MFCD00053557
≥98.5%
98.5
99%
184.65
2-Amino-6-chlorobenzothiazole

Specifications

184.65
50g
197° to 201°C
99%
C7H5ClN2S
98.5
Authentic
≥98.5%
Crystalline Powder
Off-white to light beige
100.0
95-24-9
MFCD00053557
Glass bottle
2-Amino-6-chlorobenzothiazole

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.