missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-chlorobenzoic acid, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 300500100

 View more versions of this product

Catalog No. AC300500100


 

Specifications Description & Specifications

Specifications

2-Amino-6-chlorobenzoic acid
Glass bottle
Authentic
100.0
Light yellow to beige
Crystalline Powder
99%
171.58
6-Chloroanthranilic acid
99%
2148-56-3
98.5
143° to 147°C
10g
≥98.5%
C7H6ClNO2
MFCD00051530

Chemical Identifiers

2148-56-3
100.0
C7H6ClNO2
MFCD00051530
2-Amino-6-chlorobenzoic acid, 99%
98.5
99%
171.58
6-Chloroanthranilic acid
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 218-416-8

TSCA : TSCA