missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-chloro-4-pyrimidinol Monohydrate 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 103430050

 View more versions of this product

Catalog No. AC103430050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94% (HPLC)
145.55
Light yellow
94.0
95%
C4H4ClN3O·xH2O
206658-81-3
Authentic
Powder
5g
100.0
2-Amino-6-chloro-4-pyrimidinol hydrate
MFCD00149407
95%
Glass bottle

Chemical Identifiers

206658-81-3
100.0
C4H4ClN3O·xH2O
MFCD00149407
≥94% (HPLC)
94.0
95%
145.55
2-Amino-6-chloro-4-pyrimidinol hydrate, 95%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 214-785-4

RTECSNumber : UW5995000