Please login to your online account to display your discounted pricing

2-Amino-6-chloro-4-pyrimidinol Monohydrate 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-6-chloro-4-pyrimidinol hydrate
Name Note 95%
CAS 206658-81-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94% (HPLC)
Assay Percent Range 95%
Molecular Formula C4H4ClN3O·xH2O
Formula Weight 145.55
MDL Number MFCD00149407
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC103430050

  SDS
thermo fisher scientific
103430050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94% (HPLC)
Physical Form Powder
Formula Weight 145.55
Quantity 5g
Color Light yellow
RTECS Number UW5995000
EINECS Number 214-785-4
CAS Max % 100.0
CAS Min % 94.0
Chemical Name or Material 2-Amino-6-chloro-4-pyrimidinol hydrate
Name Note 95%
MDL Number MFCD00149407
Molecular Formula C4H4ClN3O·xH2O
Assay Percent Range 95%
CAS 206658-81-3
Packaging Glass bottle
Infrared Spectrum Authentic