Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-6-chloro-4-nitrophenol, 97%

Chemical Identifiers

Chemical Name or Material 2-Amino-6-chloro-4-nitrophenol
CAS 6358-09-4
Formula Weight 188.57
MDL Number MFCD00035767
UN Number UN2811
Beilstein 2727724
Synonym 3-Chloro-2-hydroxy-5-nitroaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6676603

 
alfa aesar™
H6676603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6676606

 
alfa aesar™
H6676606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6676614

 
alfa aesar™
H6676614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 160° to 164°C
Formula Weight 188.57
Chemical Name or Material 2-Amino-6-chloro-4-nitrophenol
Physical Form Powder
Assay 97%
TSCA TSCA
MDL Number MFCD00035767
CAS 6358-09-4
UN Number UN2811
Synonym 3-Chloro-2-hydroxy-5-nitroaniline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number ST5743000
Color Orange
Quantity 1g
Beilstein 2727724
Storage Note 1 -30° to -10°C
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C