Please login to your online account to display your discounted pricing

2-Amino-6-bromopyridine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-6-bromopyridine
CAS 19798-81-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H5BrN2
Formula Weight 173.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376871000

  SDS
thermo fisher scientific
AC376871000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC376870050

  SDS
thermo fisher scientific
AC376870050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC376870250

  SDS
thermo fisher scientific
AC376870250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.01
Assay ≥97.5%
Physical Form Powder
Quantity 100g
Color Slightly yellow to light brown
Melting Point 86° to 90°C
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 98%
CAS Min % 97.5
Chemical Name or Material 2-Amino-6-bromopyridine
CAS 19798-81-3
Molecular Formula C5H5BrN2
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.