Learn More
2-Amino-6-bromopyridine, 98%, Thermo Scientific Chemicals
$137.27 - $404.00
Chemical Identifiers
CAS | 19798-81-3 |
---|---|
Molecular Formula | C5H6BrN2 |
Molecular Weight (g/mol) | 174.02 |
MDL Number | MFCD00137843 |
InChI Key | BKLJUYPLUWUEOQ-UHFFFAOYSA-O |
Synonym | 2-amino-6-bromopyridine, 6-bromo-2-pyridinamine, aminobromopyridine, 6-bromo-2-aminopyridine, 6-bromo-pyridin-2-ylamine, 2-pyridinamine, 6-bromo, 6-bromo-2-pyridylamine, 2-amino-6-bromo-pyridine, zlchem 805, pubchem7663 |
PubChem CID | 300809 |
IUPAC Name | 6-bromopyridin-2-amine |
SMILES | NC1=CC=CC(Br)=[NH+]1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2655506
|
Thermo Scientific Chemicals
H2655506 |
5 g |
Each for $137.27
|
|
|||||
AAH2655514
|
Thermo Scientific Chemicals
H2655514 |
25 g |
Each for $404.00
|
|
|||||
Description
Employed in an efficient one-pot synthesis of 7-azaindoles. Used in the synthesis of anti-HIV agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed in an efficient one-pot synthesis of 7-azaindoles. Used in the synthesis of anti-HIV agents.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
19798-81-3 | |
174.02 | |
BKLJUYPLUWUEOQ-UHFFFAOYSA-O | |
300809 | |
NC1=CC=CC(Br)=[NH+]1 |
C5H6BrN2 | |
MFCD00137843 | |
2-amino-6-bromopyridine, 6-bromo-2-pyridinamine, aminobromopyridine, 6-bromo-2-aminopyridine, 6-bromo-pyridin-2-ylamine, 2-pyridinamine, 6-bromo, 6-bromo-2-pyridylamine, 2-amino-6-bromo-pyridine, zlchem 805, pubchem7663 | |
6-bromopyridin-2-amine |
Specifications
19798-81-3 | |
C5H6BrN2 | |
5 g | |
Slightly soluble in water. | |
NC1=CC=CC(Br)=[NH+]1 | |
174.02 | |
173.01 | |
2-Amino-6-bromopyridine |
88°C to 91°C | |
MFCD00137843 | |
2-amino-6-bromopyridine, 6-bromo-2-pyridinamine, aminobromopyridine, 6-bromo-2-aminopyridine, 6-bromo-pyridin-2-ylamine, 2-pyridinamine, 6-bromo, 6-bromo-2-pyridylamine, 2-amino-6-bromo-pyridine, zlchem 805, pubchem7663 | |
BKLJUYPLUWUEOQ-UHFFFAOYSA-O | |
6-bromopyridin-2-amine | |
300809 | |
98% |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only