missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-6-bromo-3-nitropyridine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC452350010
Specifications
2-Amino-6-bromo-3-nitropyridine | |
203.0°C to 204.0°C | |
84487-04-7 | |
C5H4BrN3O2 | |
YRXZIHANXVKUHP-UHFFFAOYSA-N | |
6-bromo-3-nitropyridin-2-amine | |
12097745 | |
97% |
Glass Bottle | |
1g | |
96% min. (HPLC) | |
2-amino-3-nitro-6-bromopyridine, 2-amino-6-bromo-3-nitropyridine, 2-pyridinamine, 6-bromo-3-nitro, 2-pyridinamine,6-bromo-3-nitro, 6-amino-2-bromo-5-nitropyridine, 6-bromo-3-nitro-2-pyridinamine, 6-bromo-3-nitro-2-pyridylamine, 6-bromo-3-nitro-pyridin-2-ylamine, acmc-209puv, chempacific 38154 | |
C1=CC(=NC(=C1[N+](=O)[O-])N)Br | |
218.01 | |
218.01 |
Chemical Identifiers
84487-04-7 | |
218.01 | |
2-amino-3-nitro-6-bromopyridine, 2-amino-6-bromo-3-nitropyridine, 2-pyridinamine, 6-bromo-3-nitro, 2-pyridinamine,6-bromo-3-nitro, 6-amino-2-bromo-5-nitropyridine, 6-bromo-3-nitro-2-pyridinamine, 6-bromo-3-nitro-2-pyridylamine, 6-bromo-3-nitro-pyridin-2-ylamine, acmc-209puv, chempacific 38154 | |
6-bromo-3-nitropyridin-2-amine |
C5H4BrN3O2 | |
YRXZIHANXVKUHP-UHFFFAOYSA-N | |
12097745 | |
C1=CC(=NC(=C1[N+](=O)[O-])N)Br |
Safety and Handling
GHS Signal Word: Warning