missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-bromo-3-nitropyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 84487-04-7
Molecular Formula C5H4BrN3O2
Molecular Weight (g/mol) 218.01
InChI Key YRXZIHANXVKUHP-UHFFFAOYSA-N
Synonym 2-amino-3-nitro-6-bromopyridine, 2-amino-6-bromo-3-nitropyridine, 2-pyridinamine, 6-bromo-3-nitro, 2-pyridinamine,6-bromo-3-nitro, 6-amino-2-bromo-5-nitropyridine, 6-bromo-3-nitro-2-pyridinamine, 6-bromo-3-nitro-2-pyridylamine, 6-bromo-3-nitro-pyridin-2-ylamine, acmc-209puv, chempacific 38154
PubChem CID 12097745
IUPAC Name 6-bromo-3-nitropyridin-2-amine
SMILES C1=CC(=NC(=C1[N+](=O)[O-])N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC452350010
SDS View Documents
Thermo Fisher Scientific
AC452350010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC452350050
View Documents
Thermo Fisher Scientific
AC452350050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

84487-04-7
218.01
2-amino-3-nitro-6-bromopyridine, 2-amino-6-bromo-3-nitropyridine, 2-pyridinamine, 6-bromo-3-nitro, 2-pyridinamine,6-bromo-3-nitro, 6-amino-2-bromo-5-nitropyridine, 6-bromo-3-nitro-2-pyridinamine, 6-bromo-3-nitro-2-pyridylamine, 6-bromo-3-nitro-pyridin-2-ylamine, acmc-209puv, chempacific 38154
6-bromo-3-nitropyridin-2-amine
C5H4BrN3O2
YRXZIHANXVKUHP-UHFFFAOYSA-N
12097745
C1=CC(=NC(=C1[N+](=O)[O-])N)Br

Specifications

Glass Bottle
1g
96% min. (HPLC)
2-amino-3-nitro-6-bromopyridine, 2-amino-6-bromo-3-nitropyridine, 2-pyridinamine, 6-bromo-3-nitro, 2-pyridinamine,6-bromo-3-nitro, 6-amino-2-bromo-5-nitropyridine, 6-bromo-3-nitro-2-pyridinamine, 6-bromo-3-nitro-2-pyridylamine, 6-bromo-3-nitro-pyridin-2-ylamine, acmc-209puv, chempacific 38154
C1=CC(=NC(=C1[N+](=O)[O-])N)Br
218.01
218.01
2-Amino-6-bromo-3-nitropyridine
203.0°C to 204.0°C
84487-04-7
C5H4BrN3O2
YRXZIHANXVKUHP-UHFFFAOYSA-N
6-bromo-3-nitropyridin-2-amine
12097745
97%
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS