2-Amino-6-bromo-3-nitropyridine, 97%, ACROS Organics™
Manufacturer:  Thermo Fisher Scientific AC452350050
 View more versions of this product
Catalog No. AC452350050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Specifications
2-Amino-6-bromo-3-nitropyridine
5g
96% min. (HPLC)
2-amino-3-nitro-6-bromopyridine, 2-amino-6-bromo-3-nitropyridine, 2-pyridinamine, 6-bromo-3-nitro, 2-pyridinamine,6-bromo-3-nitro, 6-amino-2-bromo-5-nitropyridine, 6-bromo-3-nitro-2-pyridinamine, 6-bromo-3-nitro-2-pyridylamine, 6-bromo-3-nitro-pyridin-2-ylamine, acmc-209puv, chempacific 38154
C1=CC(=NC(=C1[N+](=O)[O-])N)Br
218.01
218.01
203.0°C to 204.0°C
84487-04-7
C5H4BrN3O2
YRXZIHANXVKUHP-UHFFFAOYSA-N
6-bromo-3-nitropyridin-2-amine
12097745
97%
Chemical Identifiers
84487-04-7
218.01
2-amino-3-nitro-6-bromopyridine, 2-amino-6-bromo-3-nitropyridine, 2-pyridinamine, 6-bromo-3-nitro, 2-pyridinamine,6-bromo-3-nitro, 6-amino-2-bromo-5-nitropyridine, 6-bromo-3-nitro-2-pyridinamine, 6-bromo-3-nitro-2-pyridylamine, 6-bromo-3-nitro-pyridin-2-ylamine, acmc-209puv, chempacific 38154
6-bromo-3-nitropyridin-2-amine
C5H4BrN3O2
YRXZIHANXVKUHP-UHFFFAOYSA-N
12097745
C1=CC(=NC(=C1[N+](=O)[O-])N)Br
Safety and Handling
Safety and Handling
GHS Signal Word: Warning