Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-(trifluoromethyl)pyridine, 97%

Chemical Identifiers

Chemical Name or Material 2-Amino-5-(trifluoromethyl)pyridine
CAS 74784-70-6
Molecular Formula C6H5F3N2
Formula Weight 162.11
MDL Number MFCD00042164
UN Number UN2811
Beilstein 4800784
Synonym 5-Trifluoromethyl-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2001103

  SDS
alfa aesar™
L2001103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL20011MD

  SDS
alfa aesar™
L20011MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.11
Chemical Name or Material 2-Amino-5-(trifluoromethyl)pyridine
Assay 97%
Molecular Formula C6H5F3N2
MDL Number MFCD00042164
CAS 74784-70-6
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 45° to 49°C
Quantity 1g
Beilstein 4800784
Synonym 5-Trifluoromethyl-2-pyridinamine
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.