missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-5-(trifluoromethyl)benzoic acid, 95%, Thermo Scientific Chemicals
$233.55 - $760.08
Chemical Identifiers
CAS | 83265-53-6 |
---|---|
Molecular Formula | C8H6F3NO2 |
Molecular Weight (g/mol) | 205.136 |
MDL Number | MFCD01569537 |
InChI Key | GLCQUPLYYXSPQB-UHFFFAOYSA-N |
Synonym | 2-amino-5-trifluoromethyl benzoic acid, 2-amino-5-trifluoromethyl-benzoic acid, 5-trifluoromethylanthranilic acid, 5-trifluoromethyl anthranilic acid, benzoic acid, 2-amino-5-trifluoromethyl, 2-amino-5-trifluoromethyl-benzoicacid, pubchem12904, 4-amino-3-carboxybenzotrifluoride, 2-carboxy-4-trifluoromethyl aniline, 2-amino-5-trifluoromethyl benzoicacid |
PubChem CID | 10465535 |
IUPAC Name | 2-amino-5-(trifluoromethyl)benzoic acid |
SMILES | C1=CC(=C(C=C1C(F)(F)F)C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH64141MD
|
Thermo Scientific Chemicals
H64141MD |
250 mg |
N/A
|
|
|||||
AAH6414103
|
Thermo Scientific Chemicals
H6414103 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
83265-53-6 | |
205.136 | |
GLCQUPLYYXSPQB-UHFFFAOYSA-N | |
10465535 | |
C1=CC(=C(C=C1C(F)(F)F)C(=O)O)N |
C8H6F3NO2 | |
MFCD01569537 | |
2-amino-5-trifluoromethyl benzoic acid, 2-amino-5-trifluoromethyl-benzoic acid, 5-trifluoromethylanthranilic acid, 5-trifluoromethyl anthranilic acid, benzoic acid, 2-amino-5-trifluoromethyl, 2-amino-5-trifluoromethyl-benzoicacid, pubchem12904, 4-amino-3-carboxybenzotrifluoride, 2-carboxy-4-trifluoromethyl aniline, 2-amino-5-trifluoromethyl benzoicacid | |
2-amino-5-(trifluoromethyl)benzoic acid |
Specifications
83265-53-6 | |
>110°C (230°F) | |
MFCD01569537 | |
2-amino-5-trifluoromethyl benzoic acid, 2-amino-5-trifluoromethyl-benzoic acid, 5-trifluoromethylanthranilic acid, 5-trifluoromethyl anthranilic acid, benzoic acid, 2-amino-5-trifluoromethyl, 2-amino-5-trifluoromethyl-benzoicacid, pubchem12904, 4-amino-3-carboxybenzotrifluoride, 2-carboxy-4-trifluoromethyl aniline, 2-amino-5-trifluoromethyl benzoicacid | |
C1=CC(=C(C=C1C(F)(F)F)C(=O)O)N | |
205.136 | |
205.14 | |
Solid |
141°C to 146°C (decomposition) | |
C8H6F3NO2 | |
250 mg | |
GLCQUPLYYXSPQB-UHFFFAOYSA-N | |
2-amino-5-(trifluoromethyl)benzoic acid | |
10465535 | |
95% | |
2-Amino-5-(trifluoromethyl)benzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only