Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole, 98%

$35.41 - $420.39

Chemical Identifiers

Chemical Name or Material 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole
CAS 10444-89-0
Molecular Formula C3H2F3N3S
Formula Weight 169.13
MDL Number MFCD00003109
Beilstein 513140
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1706103

  SDS
alfa aesar™
A1706103
1g Each for $35.41

AAA1706106

  SDS
alfa aesar™
A1706106
5g Each for $109.38

AAA1706114

  SDS
alfa aesar™
A1706114
25g Each for $420.39
Description & Specifications

Specifications

Formula Weight 169.13
Quantity 1g
Melting Point 222° to 227°C
Health Hazard 1 Warning
Assay 98%
Molecular Formula C3H2F3N3S
TSCA TSCA
Chemical Name or Material 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole
CAS 10444-89-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 513140
MDL Number MFCD00003109
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.