Please login to your online account to display your discounted pricing

2-Amino-5-trifluoromethyl-1,3,4-thiadiazole, 97%, ACROS Organics™

$406.40 - $406.40

Chemical Identifiers

Chemical Name or Material 2-Amino-5-trifluoromethyl-1, 3, 4-thiadiazole
Name Note 97%
CAS 10444-89-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C3 H2 F3 N3 S
Formula Weight 169.13
MDL Number MFCD00003109
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC188510250

  SDS
thermo fisher scientific
AC188510250
Glass bottle 25g Each for $406.40

AC188510010

  SDS
thermo fisher scientific
AC188510010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 169.13
Quantity 25g
Color White to off-white
Melting Point 220° to 225°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Amino-5-trifluoromethyl-1, 3, 4-thiadiazole
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C3 H2 F3 N3 S
MDL Number MFCD00003109
CAS 10444-89-0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic