Learn More
2-Amino-5-picoline, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 104390100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Amino-5-picoline | |
1603-41-4 | |
100.0 | |
White to Yellow | |
Authentic | |
Glass bottle | |
MFCD00006328 | |
22, V,9, 289 | |
Solubility in water: freely soluble. | |
CC1=CN=C(C=C1)N | |
108.14 | |
108.14 | |
Crystalline Powder, Crystals or Flakes |
99% | |
98.5 | |
80.0°C to 81.0°C | |
227.0°C | |
98.5% min. (GC) | |
C6H8N2 | |
10 g | |
2-amino-5-methylpyridine, 2-amino-5-picoline, 6-amino-3-picoline, 2-pyridinamine, 5-methyl, 5-methylpyridin-2-ylamine, 3-picoline, 6-amino, 5-methyl-2-pyridinamine, 5-methyl-2-pyridylamine, 5-methyl-2-aminopyridine, 2-amino-5-methyl pyridine | |
CMBSSVKZOPZBKW-UHFFFAOYSA-N | |
5-methylpyridin-2-amine | |
15348 | |
99% |
Chemical Identifiers
1603-41-4 | |
108.14 | |
CMBSSVKZOPZBKW-UHFFFAOYSA-N | |
15348 | |
CC1=CN=C(C=C1)N |
C6H8N2 | |
MFCD00006328 | |
2-amino-5-methylpyridine, 2-amino-5-picoline, 6-amino-3-picoline, 2-pyridinamine, 5-methyl, 5-methylpyridin-2-ylamine, 3-picoline, 6-amino, 5-methyl-2-pyridinamine, 5-methyl-2-pyridylamine, 5-methyl-2-aminopyridine, 2-amino-5-methyl pyridine | |
5-methylpyridin-2-amine |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Toxic in contact with skin.
Toxic if inhaled.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 216-503-5
RTECSNumber : TJ5141000
TSCA : TSCA
RUO – Research Use Only