Please login to your online account to display your discounted pricing

2-Amino-5-phenyl-3-furonitrile, 95%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  AW01254EA

 View more versions of this product

Catalog No. AW01254EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 184.2
Quantity 10g
Assay 95%
CAS Max % 100.0
CAS 14742-32-6
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 95%
CAS Min % 94.5
Chemical Name or Material 2-Amino-5-phenyl-3-furonitrile
Molecular Formula C11H8N2O
MDL Number MFCD00463472
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 2-Amino-5-phenyl-3-furonitrile
CAS 14742-32-6
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H8N2O
Formula Weight 184.2
MDL Number MFCD00463472
Accessories

GHS Signal Word: Warning

SDS & Safety Documents