Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-nitropyrimidine, 97%

$164.16 - $164.16

Chemical Identifiers

Chemical Name or Material 2-Amino-5-nitropyrimidine
CAS 3073-77-6
Formula Weight 140.10g/mol
MDL Number MFCD00006103
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4149504

 
alfa aesar™
41495-04
2g Each for $164.16
Description & Specifications

Specifications

Melting Point 235° to 238°C
Quantity 2g
Chemical Name or Material 2-Amino-5-nitropyrimidine
CAS 3073-77-6
Physical Form Powder
Assay 97%
Formula Weight 140.10g/mol
MDL Number MFCD00006103
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.