missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-5-nitropyridine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 104230250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Amino-5-nitropyridine | |
98.5 | |
186.0°C to 190.0°C | |
Authentic | |
Glass bottle | |
NH2(C5H3N)NO2 | |
25 g | |
2-amino-5-nitropyridine, 2-pyridinamine, 5-nitro, 5-nitro-2-aminopyridine, 5-nitro-2-pyridinamine, 2-amino-5-nitro pyridine, pyridine, 2-amino-5-nitro, 5-nitro-pyridin-2-ylamine, 5-nitro-pyridin-2-yl amine, 5-nitro-2-pyridylamine, 5-nitropyridin-2-amin | |
UGSBCCAHDVCHGI-UHFFFAOYSA-N | |
5-nitropyridin-2-amine | |
77888 | |
99% |
4214-76-0 | |
100.0 | |
Yellow | |
98.5% min. (HPLC) | |
C5H5N3O2 | |
MFCD00006325 | |
22,I,631 | |
Solubility in water: 1.6g/L water (13°C). | |
NC1=CC=C(C=N1)[N+]([O-])=O | |
139.11 | |
139.11 | |
Crystalline Powder |
Chemical Identifiers
4214-76-0 | |
139.11 | |
UGSBCCAHDVCHGI-UHFFFAOYSA-N | |
77888 | |
NC1=CC=C(C=N1)[N+]([O-])=O |
C5H5N3O2 | |
MFCD00006325 | |
2-amino-5-nitropyridine, 2-pyridinamine, 5-nitro, 5-nitro-2-aminopyridine, 5-nitro-2-pyridinamine, 2-amino-5-nitro pyridine, pyridine, 2-amino-5-nitro, 5-nitro-pyridin-2-ylamine, 5-nitro-pyridin-2-yl amine, 5-nitro-2-pyridylamine, 5-nitropyridin-2-amin | |
5-nitropyridin-2-amine |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 224-145-6
RUO – Research Use Only