missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-nitropyridine, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1135236

 View more versions of this product

Catalog No. AAA1135236



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-5-nitropyridine
185°C to 190°C
C5H5N3O2
500 g
Light sensitive
UGSBCCAHDVCHGI-UHFFFAOYSA-N
5-nitropyridin-2-amine
77888
98%
4214-76-0
224°C (435°F)
MFCD00006325
120353
2-amino-5-nitropyridine, 2-pyridinamine, 5-nitro, 5-nitro-2-aminopyridine, 5-nitro-2-pyridinamine, 2-amino-5-nitro pyridine, pyridine, 2-amino-5-nitro, 5-nitro-pyridin-2-ylamine, 5-nitro-pyridin-2-yl amine, 5-nitro-2-pyridylamine, 5-nitropyridin-2-amin
NC1=CC=C(C=N1)[N+]([O-])=O
139.11
139.11

Chemical Identifiers

4214-76-0
139.11
UGSBCCAHDVCHGI-UHFFFAOYSA-N
77888
NC1=CC=C(C=N1)[N+]([O-])=O
C5H5N3O2
MFCD00006325
2-amino-5-nitropyridine, 2-pyridinamine, 5-nitro, 5-nitro-2-aminopyridine, 5-nitro-2-pyridinamine, 2-amino-5-nitro pyridine, pyridine, 2-amino-5-nitro, 5-nitro-pyridin-2-ylamine, 5-nitro-pyridin-2-yl amine, 5-nitro-2-pyridylamine, 5-nitropyridin-2-amin
5-nitropyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 224-145-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only