missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-nitrophenol 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC400900250

 View more versions of this product

Catalog No. AC400900250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
Crystalline Powder
Rust brown to brown
94.0
121-88-0
2-Amino-5-nitrophenol
MFCD00007692
2-Hydroxy-4-nitroaniline
154.13
25g
100.0
Glass bottle
C6H6N2O3
95%
95%
Authentic

Chemical Identifiers

121-88-0
100.0
C6H6N2O3
MFCD00007692
2-Amino-5-nitrophenol, 95%
94.0
95%
154.13
2-Hydroxy-4-nitroaniline
≥94%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing genetic defects.
Causes serious eye irritation.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

CANCER SUSPECTED AGENT

EINECSNumber : 204-503-8

RTECSNumber : SJ6302500

TSCA : TSCA