missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-nitrobenzotrifluoride, 98%, ACROS Organics™

Manufacturer:  Acros Organics 188230050

 View more versions of this product

Catalog No. AC188230050


 

Specifications

Specifications

2-Amino-5-nitrobenzotrifluoride
121-01-7
100.0
Glass bottle
Yellow through Brown
98%
C7H5F3N2O2
MFCD00007365
HOTZLWVITTVZGY-UHFFFAOYSA-N
4-nitro-2-(trifluoromethyl)aniline
67128
Powder
98%
97.5
Authentic
90°C to 93°C
≥97.5%
5g
H2NC6H3(NO2)CF3
4-Nitro-α, α, α-trifluoro-o-toluidine, 4-Nitro-α, α, α-trifluoro-o-toluidine
C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)N
206.124
206.12

Chemical Identifiers

121-01-7
206.124
HOTZLWVITTVZGY-UHFFFAOYSA-N
67128
C7H5F3N2O2
MFCD00007365
4-Nitro-α, α, α-trifluoro-o-toluidine, 4-Nitro-α, α, α-trifluoro-o-toluidine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-443-2

TSCA : TSCA

SDS Documents