missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-nitrobenzotrifluoride, 98%, ACROS Organics™

$38.87 - $38.87

Chemical Identifiers

CAS 121-01-7
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.12
MDL Number MFCD00007365
InChI Key HOTZLWVITTVZGY-UHFFFAOYSA-N
Synonym 2-amino-5-nitro-trifluorobenzene, 2-amino-5-nitrobenzotrifluoride, 2-trifluoromethyl-4-nitroaniline, 4-nitro-2-trifluoromethyl aniline, 4-nitro-2-trifluoromethyl benzenamine, 4-nitro-a,a,a-trifluoro-o-toluidine, 4-nitro-alpha,alpha,alpha-trifluoro-o-toluidine, 5-nitro-2-aminobenzotrifluoride, a,a,a-trifluoro-4-nitro-o-toluidine, benzenamine, 4-nitro-2-trifluoromethyl
PubChem CID 67128
IUPAC Name 4-nitro-2-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC188230050
SDS View Documents
Thermo Fisher Scientific
188230050
5g
Each for $38.87
 
Description

Description

Specifications

Chemical Identifiers

121-01-7
206.12
HOTZLWVITTVZGY-UHFFFAOYSA-N
67128
C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)N
C7H5F3N2O2
MFCD00007365
2-amino-5-nitro-trifluorobenzene, 2-amino-5-nitrobenzotrifluoride, 2-trifluoromethyl-4-nitroaniline, 4-nitro-2-trifluoromethyl aniline, 4-nitro-2-trifluoromethyl benzenamine, 4-nitro-a,a,a-trifluoro-o-toluidine, 4-nitro-alpha,alpha,alpha-trifluoro-o-toluidine, 5-nitro-2-aminobenzotrifluoride, a,a,a-trifluoro-4-nitro-o-toluidine, benzenamine, 4-nitro-2-trifluoromethyl
4-nitro-2-(trifluoromethyl)aniline

Specifications

Authentic
Yellow to Brown
5g
97.5
98%
H2NC6H3(NO2)CF3
2-amino-5-nitro-trifluorobenzene, 2-amino-5-nitrobenzotrifluoride, 2-trifluoromethyl-4-nitroaniline, 4-nitro-2-trifluoromethyl aniline, 4-nitro-2-trifluoromethyl benzenamine, 4-nitro-a,a,a-trifluoro-o-toluidine, 4-nitro-alpha,alpha,alpha-trifluoro-o-toluidine, 5-nitro-2-aminobenzotrifluoride, a,a,a-trifluoro-4-nitro-o-toluidine, benzenamine, 4-nitro-2-trifluoromethyl
C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)N
206.12
206.12
98%
Glass bottle
90°C to 93°C
121-01-7
100.0
C7H5F3N2O2
MFCD00007365
HOTZLWVITTVZGY-UHFFFAOYSA-N
4-nitro-2-(trifluoromethyl)aniline
67128
Powder
2-Amino-5-nitrobenzotrifluoride, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-443-2

TSCA : TSCA

SDS
Product Certifications