Learn More
2-Amino-5-nitrobenzonitrile, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2541622
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Amino-5-nitrobenzonitrile | |
207°C to 210°C | |
MFCD00007362 | |
5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j | |
C1=CC(=C(C=C1[N+](=O)[O-])C#N)N | |
163.136 | |
163.14 |
17420-30-3 | |
C7H5N3O2 | |
100 g | |
MGCGMYPNXAFGFA-UHFFFAOYSA-N | |
2-amino-5-nitrobenzonitrile | |
28532 | |
95% |
Chemical Identifiers
17420-30-3 | |
163.136 | |
MGCGMYPNXAFGFA-UHFFFAOYSA-N | |
28532 | |
C1=CC(=C(C=C1[N+](=O)[O-])C#N)N |
C7H5N3O2 | |
MFCD00007362 | |
5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j | |
2-amino-5-nitrobenzonitrile |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 241-446-8
RTECSNumber : BX2500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only