Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-nitrobenzoic acid potassium salt, 98%

$102.99 - $1,326.86

Chemical Identifiers

Chemical Name or Material 2-Amino-5-nitrobenzoic acid potassium salt
CAS 209664-72-2
Molecular Formula C7H5KN2O4
Formula Weight 220.24
MDL Number MFCD00156119
Synonym Potassium 2-amino-5-nitrobenzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1481914

  SDS
alfa aesar™
A1481914
25g Each for $102.99

AAA1481922

  SDS
alfa aesar™
A1481922
100g Each for $318.64

AAA1481936

  SDS
alfa aesar™
A1481936
500g Each for $1,326.86
Description & Specifications

Specifications

Formula Weight 220.24
Chemical Name or Material 2-Amino-5-nitrobenzoic acid potassium salt
Melting Point >300°C
Assay 98%
Quantity 25g
Molecular Formula C7H5KN2O4
MDL Number MFCD00156119
CAS 209664-72-2
Synonym Potassium 2-amino-5-nitrobenzoate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.