missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-nitrobenzoic acid potassium salt, 98%

Manufacturer:  Alfa Aesar™ A1481936

 View more versions of this product

Catalog No. AAA1481936



Specifications

Specifications

2-Amino-5-nitrobenzoic acid potassium salt
500g
C7H5KN2O4
potassium 2-amino-5-nitrobenzoate, 2-amino-5-nitrobenzoic acid potassium salt, potassium 5-nitroanthranilate, 2-amino-5-nitrobenzoic acid potassium salt
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)[O-])N.[K+]
220.225
220.24
>300°C
209664-72-2
MFCD00156119
VUOPWTVKTDYMCB-UHFFFAOYSA-M
potassium;2-amino-5-nitrobenzoate
68494575
98%

Chemical Identifiers

209664-72-2
220.225
VUOPWTVKTDYMCB-UHFFFAOYSA-M
68494575
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)[O-])N.[K+]
C7H5KN2O4
MFCD00156119
potassium 2-amino-5-nitrobenzoate, 2-amino-5-nitrobenzoic acid potassium salt, potassium 5-nitroanthranilate, 2-amino-5-nitrobenzoic acid potassium salt
potassium;2-amino-5-nitrobenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents