Learn More
Alfa Aesar™ 2-Amino-5-nitrobenzoic acid, 98%
$73.70 - $1,116.50
Chemical Identifiers
CAS | 616-79-5 |
---|---|
Molecular Formula | C7H6N2O4 |
Molecular Weight (g/mol) | 182.135 |
MDL Number | MFCD00017039 |
InChI Key | RUCHWTKMOWXHLU-UHFFFAOYSA-N |
Synonym | 5-nitroanthranilic acid, benzoic acid, 2-amino-5-nitro, 2-amino-5-nitro-benzoic acid, 2-amino-5-nitrobenzoicacid, anthranilic acid, 5-nitro, 1-amino-2-carboxy-4-nitrobenzene, 2-amino-5-nitro-benzoicacid, pubchem14422, acmc-209mvd, 5-nitro anthranilic acid |
PubChem CID | 12032 |
ChEBI | CHEBI:61280 |
IUPAC Name | 2-amino-5-nitrobenzoic acid |
SMILES | C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1042609 |
Alfa Aesar™
A1042609 |
10g |
Each for $73.70
|
|
AAA1042618 |
Alfa Aesar™
A1042618 |
50g |
Each for $246.74
|
|
AAA1042630 |
Alfa Aesar™
A1042630 |
250g |
Each for $1,116.50
|
|
Description
Applications2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to strong oxidizing agents. Stable under ordinary conditions.
Chemical Identifiers
616-79-5 | |
182.135 | |
RUCHWTKMOWXHLU-UHFFFAOYSA-N | |
12032 | |
2-amino-5-nitrobenzoic acid |
C7H6N2O4 | |
MFCD00017039 | |
5-nitroanthranilic acid, benzoic acid, 2-amino-5-nitro, 2-amino-5-nitro-benzoic acid, 2-amino-5-nitrobenzoicacid, anthranilic acid, 5-nitro, 1-amino-2-carboxy-4-nitrobenzene, 2-amino-5-nitro-benzoicacid, pubchem14422, acmc-209mvd, 5-nitro anthranilic acid | |
CHEBI:61280 | |
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)N |
Specifications
~270°C (decomposition) | |
616-79-5 | |
MFCD00017039 | |
5-nitroanthranilic acid, benzoic acid, 2-amino-5-nitro, 2-amino-5-nitro-benzoic acid, 2-amino-5-nitrobenzoicacid, anthranilic acid, 5-nitro, 1-amino-2-carboxy-4-nitrobenzene, 2-amino-5-nitro-benzoicacid, pubchem14422, acmc-209mvd, 5-nitro anthranilic acid | |
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)N | |
182.135 | |
CHEBI:61280 | |
98% |
10g | |
C7H6N2O4 | |
646219 | |
RUCHWTKMOWXHLU-UHFFFAOYSA-N | |
2-amino-5-nitrobenzoic acid | |
12032 | |
182.14 | |
2-Amino-5-nitrobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA